Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014453.3 | NP_055268.1 | 5 | 684 | ||
NM_198426.2 | NP_940818.1 | 5 | 686 | |||
XM_005258746.2 | XP_005258803.1 | 5 | 1184 | |||
XM_005258747.3 | XP_005258804.1 | 5 | 1307 | |||
GenBank mRNA | AF042384.1 | - | 5 | 652 | ||
AK311974.1 | - | 5 | 572 | |||
AY364248.1 | - | 4 | 523 | |||
BC002502.2 | - | 5 | 656 | 3051024 | ||
BG546479.1 | - | 5 | 684 | |||
BI457978.1 | - | 5 | 677 | |||
BT007298.1 | - | 4 | 523 | |||
BU147101.1 | - | 3 | 331 | |||
CR457002.1 | - | 4 | 523 |