Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029882.3 | NP_001025053.1 | 6 | 2303 | ||
XM_005245848.3 | XP_005245905.1 | 7 | 2538 | |||
XM_005245849.3 | XP_005245906.1 | 7 | 2643 | |||
XM_005245850.3 | XP_005245907.1 | 8 | 2663 | |||
XM_005245851.3 | XP_005245908.1 | 7 | 2535 | |||
XM_005245852.3 | XP_005245909.1 | 6 | 2312 | |||
XM_011541255.1 | XP_011539557.1 | 7 | 2217 | |||
XM_011541256.2 | XP_011539558.1 | 7 | 2657 | |||
XM_011541257.2 | XP_011539559.1 | 6 | 3412 |