Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002755.2 | NP_001002755.1 | 6 | 736 | ||
NM_001002756.2 | NP_001002756.1 | 5 | 690 | |||
NM_015700.3 | NP_056515.2 | 6 | 826 | |||
NR_045631.1 | 5 | 600 | ||||
NR_045632.1 | 5 | 600 | ||||
XM_017003808.1 | XP_016859297.1 | 6 | 539 | |||
GenBank mRNA | AF132967.1 | - | 6 | 538 | ||
AJ132584.1 | - | 6 | 614 | |||
AK300700.1 | - | 6 | 545 | |||
AK308567.1 | - | 5 | 407 | |||
AK314004.1 | - | 6 | 569 | |||
AY286306.1 | - | 6 | 583 | |||
AY286307.1 | - | 6 | 673 | |||
AY335194.1 | - | 6 | 729 | |||
BC113692.1 | - | 6 | 698 | 8322744 | ||
BC113694.1 | - | 6 | 698 | 8322746 | ||
BG385940.1 | - | 5 | 488 | |||
BU178674.1 | - | 5 | 432 | |||
BU507991.1 | - | 6 | 541 | |||
BX538347.1 | - | 6 | 601 |