Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002755.2 | NP_001002755.1 | 7 | 855 | ||
NM_001002756.2 | NP_001002756.1 | 6 | 809 | |||
NM_015700.3 | NP_056515.2 | 7 | 945 | |||
NR_045631.1 | 6 | 719 | ||||
XM_017003808.1 | XP_016859297.1 | 7 | 658 | |||
GenBank mRNA | AF132967.1 | - | 7 | 657 | ||
AJ132584.1 | - | 7 | 733 | |||
AK300700.1 | - | 7 | 664 | |||
AK314004.1 | - | 7 | 688 | |||
AY286306.1 | - | 7 | 702 | |||
AY286307.1 | - | 7 | 792 | |||
AY335194.1 | - | 7 | 848 | |||
BC113692.1 | - | 7 | 817 | 8322744 | ||
BC113694.1 | - | 7 | 817 | 8322746 | ||
BG385940.1 | - | 6 | 606 | |||
BU178674.1 | - | 6 | 551 | |||
BU507991.1 | - | 7 | 660 | |||
BX538347.1 | - | 7 | 720 |