Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199492.1 | NP_001186421.1 | 3 | 435 | ||
NM_014456.4 | NP_055271.2 | 3 | 477 | |||
NM_145341.3 | NP_663314.1 | 4 | 561 | |||
GenBank mRNA | AK223623.1 | - | 3 | 351 | ||
AK296753.1 | - | 3 | 302 | |||
AK312540.1 | - | 3 | 373 | |||
AK315295.1 | - | 4 | 439 | |||
BC015036.2 | - | 3 | 341 | |||
BC026104.2 | - | 3 | 344 | 4798293 | ||
BC031049.1 | - | 3 | 344 | 5277621 | ||
BI910668.1 | - | 3 | 439 | |||
BX537500.1 | - | 2 | 3425 | |||
BX647834.1 | - | 3 | 321 | |||
DB241608.1 | - | 3 | 478 | |||
U96628.1 | - | 3 | 275 |