Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199492.1 | NP_001186421.1 | 9, 10 | 1336 | ||
NM_014456.4 | NP_055271.2 | 9, 10 | 1378 | |||
NM_145341.3 | NP_663314.1 | 10, 11 | 1462 | |||
GenBank mRNA | AK223623.1 | - | 9, 10 | 1252 | ||
AK296753.1 | - | 9, 10 | 1203 | |||
AK312540.1 | - | 9, 10 | 1274 | |||
AK315295.1 | - | 10, 11 | 1340 | |||
BC026104.2 | - | 9, 10 | 1245 | 4798293 | ||
BC031049.1 | - | 9, 10 | 1245 | 5277621 | ||
BX537500.1 | - | 7, 8 | 5251 | |||
BX647834.1 | - | 9, 10 | 1220 | |||
U96628.1 | - | 9, 10 | 1176 |