Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014460.3 | NP_055275.1 | 3 | 580 | ||
GenBank mRNA | AB027011.1 | - | 2 | 303 | ||
AK313407.1 | - | 3 | 537 | |||
AL834417.1 | - | 3 | 465 | |||
BC067113.1 | - | 3 | 526 | 5759182 | ||
CR456457.1 | - | 2 | 315 |