Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021950.3 | NP_068769.2 | 5 | 550 | ||
NM_152866.2 | NP_690605.1 | 6 | 813 | |||
GenBank mRNA | AK225630.1 | - | 5 | 512 | ||
AK292168.1 | - | 5 | 686 | |||
BC002807.2 | - | 5 | 500 | 3634040 | ||
CD691924.1 | - | 6 | 573 | |||
X07203.1 | - | 5 | 487 | |||
X12530.1 | - | 5 | 490 |