Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289080.1 | NP_001276009.1 | 8 | 1392 | ||
NM_001289081.1 | NP_001276010.1 | 7 | 1255 | |||
NM_014461.3 | NP_055276.1 | 8 | 1401 | |||
XM_005265058.3 | XP_005265115.1 | 9 | 1524 | |||
XM_011533590.2 | XP_011531892.1 | 8 | 1048 | |||
XM_011533591.2 | XP_011531893.1 | 7 | 1363 | |||
XM_017006171.1 | XP_016861660.1 | 13 | 1951 | |||
XM_017006172.1 | XP_016861661.1 | 8 | 1526 | |||
XM_017006173.1 | XP_016861662.1 | 7 | 1032 | |||
XM_017006174.1 | XP_016861663.1 | 7 | 1015 | |||
XM_017006175.1 | XP_016861664.1 | 5 | 689 | |||
XM_017006176.1 | XP_016861665.1 | 6 | 833 | |||
XM_017006177.1 | XP_016861666.1 | 8 | 1140 | |||
GenBank mRNA | AB003592.1 | - | 8 | 1114 | ||
AK294786.1 | - | 7 | 1255 | |||
AK314220.1 | - | 8 | 1115 | |||
BC113118.1 | - | 8 | 1027 | 40036214 |