Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014464.3 | NP_055279.3 | 1 | 280 | ||
XM_006715062.2 | XP_006715125.1 | 2 | 263 | |||
XM_011514497.2 | XP_011512799.1 | 1 | 262 | |||
XM_017010745.1 | XP_016866234.1 | 1 | 244 | |||
GenBank mRNA | AB022277.1 | - | 1 | 135 | ||
AF195116.1 | - | 1 | 134 | |||
AF195117.1 | - | 1 | 134 | |||
AK312918.1 | - | 1 | 244 | |||
BC056235.1 | - | 1 | 244 | |||
BC070278.1 | - | 1 | 280 | 4607306 |