Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014467.2 | NP_055282.1 | 7 | 1192 | ||
GenBank mRNA | AF060567.1 | - | 7 | 1130 | ||
AF393649.1 | - | 7 | 1051 | |||
AK075462.1 | - | 7 | 1140 | |||
BC020733.1 | - | 7 | 967 | 4769946 |