Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPDL3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009568.2 | NP_001009568.1 | 3, 4 | 600 | ||
NM_001304579.1 | NP_001291508.1 | 3, 4 | 671 | |||
NM_014474.3 | NP_055289.2 | 3, 4 | 600 | |||
XM_011541259.2 | XP_011539561.1 | 4, 5 | 690 | |||
XM_011541260.2 | XP_011539562.1 | 4, 5 | 1325 | |||
GenBank mRNA | AK293566.1 | - | 3, 4 | 597 | ||
AK301349.1 | - | 3, 4 | 555 | |||
AK316500.1 | - | 3, 4 | 526 | |||
BC014444.1 | - | 3, 4 | 517 | 4865268 | ||
CB160850.1 | - | 3, 4 | 562 | |||
DC370191.1 | - | 3, 4 | 599 | |||
Y08134.1 | - | 3, 4 | 492 |