Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMPDL3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009568.2 | NP_001009568.1 | 5 | 797 | ||
NM_001304579.1 | NP_001291508.1 | 5 | 868 | |||
NM_014474.3 | NP_055289.2 | 5 | 797 | |||
XM_011541259.2 | XP_011539561.1 | 6 | 887 | |||
XM_011541260.2 | XP_011539562.1 | 6 | 1522 | |||
GenBank mRNA | AK293489.1 | - | 4 | 499 | ||
AK293566.1 | - | 5 | 794 | |||
AK301349.1 | - | 5 | 752 | |||
AK316500.1 | - | 5 | 723 | |||
BC014444.1 | - | 5 | 714 | 4865268 | ||
Y08134.1 | - | 5 | 689 |