Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26140
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
4
557
NM_001256235.1
NP_001243164.1
5
498
NM_001256236.1
NP_001243165.1
5
740
NM_001256237.1
NP_001243166.1
4
557
NM_001256238.1
NP_001243167.1
4
471
NM_001256239.1
NP_001243168.1
3
487
NM_001256240.1
NP_001243169.1
3
487
NM_001283038.1
NP_001269967.1
4
557
NM_001283039.1
NP_001269968.1
4
510
NM_014489.3
NP_055304.1
4
670
NR_027016.2
2
312
NR_027017.2
5
620
NR_027018.2
5
509
NR_045923.1
3
382
NR_045927.1
3
382
NR_045929.1
3
440
NR_104270.1
3
382
NR_104271.1
5
642
NR_104272.1
2
312
XM_006718181.3
XP_006718244.1
3
1148
XM_006718183.3
XP_006718246.1
5
565
XM_006718185.2
XP_006718248.1
4
457
XM_006718186.2
XP_006718249.1
5
617
XM_006718190.3
XP_006718253.1
4
773
XM_006718191.3
XP_006718254.1
4
773
XM_006718192.1
XP_006718255.1
6
691
XM_006718193.2
XP_006718256.1
5
498
XM_011519990.2
XP_011518292.2
5
1024
XM_011519991.2
XP_011518293.2
5
1027
XM_011519992.1
XP_011518294.1
6
796
XM_011519993.2
XP_011518295.2
5
1043
XM_011519994.2
XP_011518296.2
5
1043
XM_011519995.2
XP_011518297.1
5
947
XM_011519996.1
XP_011518298.1
6
748
XM_011519997.1
XP_011518299.1
6
1089
XM_011519998.2
XP_011518300.1
6
767
XM_011519999.1
XP_011518301.1
6
726
XM_011520000.1
XP_011518302.1
5
640
XM_011520001.1
XP_011518303.1
4
432
XM_011520002.1
XP_011518304.1
4
432
XM_011520003.2
XP_011518305.1
5
956
XM_011520004.2
XP_011518306.1
5
631
XM_011520005.2
XP_011518307.1
4
528
XM_011520006.2
XP_011518308.1
5
912
XM_011520007.2
XP_011518309.1
6
681
XM_017017556.1
XP_016873045.1
4
432
XM_017017557.1
XP_016873046.1
4
580
XM_017017558.1
XP_016873047.1
4
754
XM_017017559.1
XP_016873048.1
5
617
XM_017017560.1
XP_016873049.1
5
617
GenBank mRNA
AF159615.1
-
4
672
AK292181.1
-
5
634
AK295202.1
-
5
509
AK301991.1
-
5
590
AK307359.1
-
3
299
AK308525.1
-
3
293
AL049261.1
-
2
922
AL096753.1
-
5
498
AY373030.1
-
2
361
BC009930.2
-
4
444
3506614
BG707484.1
-
3
276
BI759334.1
-
3
379
BI768071.1
-
3
444
BI771208.1
-
2
273
BI772237.1
-
4
450
BI911028.1
-
4
586
BI911526.1
-
5
531
BX340489.2
-
3
424
BX356273.2
-
4
464
BX379511.2
-
4
439
BX448155.2
-
4
416
CB988453.1
-
2
339
CD675009.1
-
1
23
CD675113.1
-
3
275
CX872317.1
-
2
240
KU178608.1
-
2
361
KU178609.1
-
3
446
KU178610.1
-
3
446
KU178611.1
-
2
361
Back To Top
More Information
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline