Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002139.3 | NP_002130.2 | 5 | 734 | ||
NR_028476.1 | 4 | 562 | ||||
NR_028477.1 | 5 | 769 | ||||
GenBank mRNA | AK091520.1 | - | 4 | 508 | ||
AK096015.1 | - | 2 | 640 | |||
AK304573.1 | - | 5 | 714 | |||
AK309911.1 | - | 5 | 689 | |||
AY464692.1 | - | 4 | 524 | |||
BC006550.2 | - | 5 | 670 | 3010123 | ||
BC007435.2 | - | 5 | 670 | 3010123 | ||
BX647131.1 | - | 4 | 505 | |||
DA925805.1 | - | 4 | 507 | |||
Z23064.1 | - | 4 | 535 |