Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNRC6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014494.2 | NP_055309.2 | 19 | 4839 | ||
XM_005255254.3 | XP_005255311.1 | 18 | 4682 | |||
XM_005255257.4 | XP_005255314.1 | 18 | 4806 | |||
XM_017023143.1 | XP_016878632.1 | 18 | 4783 | |||
XM_017023144.1 | XP_016878633.1 | 17 | 4750 | |||
XM_017023145.1 | XP_016878634.1 | 18 | 4716 | |||
XM_017023146.1 | XP_016878635.1 | 18 | 4848 | |||
XM_017023147.1 | XP_016878636.1 | 18 | 4876 | |||
XM_017023148.1 | XP_016878637.1 | 17 | 4603 | |||
XM_017023149.1 | XP_016878638.1 | 16 | 4600 | |||
XM_017023150.1 | XP_016878639.1 | 17 | 4565 | |||
XM_017023151.1 | XP_016878640.1 | 16 | 4454 | |||
XM_017023152.1 | XP_016878641.1 | 17 | 4355 | |||
XM_017023153.1 | XP_016878642.1 | 18 | 4806 | |||
XM_017023154.1 | XP_016878643.1 | 16 | 4697 | |||
GenBank mRNA | AB040893.1 | - | 14 | 3039 | ||
AJ492221.1 | - | 18 | 4725 | |||
AK001773.1 | - | 2 | 164 | |||
AK025696.1 | - | 12 | 1396 | |||
AK094542.1 | - | 6 | 1421 | |||
AL713750.1 | - | 6 | 1592 | |||
AL833757.1 | - | 13 | 2000 | |||
AY035864.1 | - | 16 | 4373 | |||
BC024324.1 | - | 2 | 185 |