Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291737.1 | NP_001278666.1 | 3 | 489 | ||
NM_001291738.1 | NP_001278667.1 | 3 | 344 | |||
NM_001291739.1 | NP_001278668.1 | 2 | 423 | |||
NM_013230.3 | NP_037362.1 | 3 | 562 | |||
NR_117089.1 | 2 | 312 | ||||
NR_117090.1 | 2 | 385 | ||||
GenBank mRNA | AK026603.1 | - | 2 | 211 | ||
AK125531.1 | - | 3 | 423 | |||
AU125939.1 | - | 3 | 489 | |||
BC064619.1 | - | 2 | 189 | 5591617 | ||
BG755979.1 | - | 3 | 563 | |||
BT007404.1 | - | 2 | 122 | |||
DA253943.1 | - | 2 | 247 | |||
DA378105.1 | - | 3 | 344 | |||
DA518957.1 | - | 2 | 423 | |||
DA745135.1 | - | 2 | 196 | |||
DA753855.1 | - | 3 | 323 | |||
L33930.1 | - | 2 | 178 | |||
M58664.1 | - | 2 | 178 | |||
X69397.1 | - | 2 | 181 |