Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324218.1 | NP_001311147.1 | 3 | 1147 | ||
NM_001324221.1 | NP_001311150.1 | 3 | 1053 | |||
NM_001324225.1 | NP_001311154.1 | 4 | 1132 | |||
NM_014499.3 | NP_055314.1 | 4 | 1196 | |||
NM_198333.2 | NP_938147.1 | 2 | 1004 | |||
GenBank mRNA | AK292138.1 | - | 4 | 1134 | ||
AK313218.1 | - | 2 | 945 | |||
BC051875.1 | - | 4 | 1107 | 5212240 | ||
BC095498.1 | - | 1 | 832 | 6971933 |