Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014502.4 | NP_055317.1 | 2, 3 | 368 | ||
GenBank mRNA | AJ131186.1 | - | 2, 3 | 368 | ||
AL555169.3 | - | 2, 3 | 379 | |||
BC008719.2 | - | 2, 3 | 363 | 2961655 | ||
BC018665.2 | - | 2, 3 | 363 | 2961655 | ||
BC018698.2 | - | 2, 3 | 363 | 2961655 |