Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287060.1 | NP_001273989.1 | 8, 9 | 1691 | ||
NM_001287061.1 | NP_001273990.1 | 7, 8 | 1031 | |||
NM_001287062.1 | NP_001273991.1 | 8, 9 | 1167 | |||
NM_014504.2 | NP_055319.1 | 7, 8 | 972 | |||
GenBank mRNA | AJ250042.1 | - | 7, 8 | 895 | ||
AK001702.1 | - | 4, 5 | 476 | |||
AK056864.1 | - | 8, 9 | 1618 | |||
AK290678.1 | - | 8, 9 | 1090 | |||
AK303006.1 | - | 7, 8 | 1028 | |||
BC015330.1 | - | 7, 8 | 918 | 4420279 | ||
BT007107.1 | - | 6, 7 | 818 | |||
DQ230533.1 | - | 6, 7 | 818 | |||
DQ230534.1 | - | 7, 8 | 938 | |||
DQ230535.1 | - | 4, 5 | 566 |