Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242.4 | NP_001233.1 | 3 | 648 | ||
XM_011521042.2 | XP_011519344.1 | 2 | 812 | |||
XM_017020232.1 | XP_016875721.1 | 3 | 666 | |||
XM_017020233.1 | XP_016875722.1 | 2 | 824 | |||
XM_017020234.1 | XP_016875723.1 | 3 | 666 | |||
GenBank mRNA | AK315732.1 | - | 3 | 667 | ||
BC012160.1 | - | 3 | 634 | 4575359 | ||
M63928.1 | - | 3 | 537 |