Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014573.2 | NP_055388.2 | 1 | 227 | ||
XM_005257965.3 | XP_005258022.1 | 1 | 281 | |||
GenBank mRNA | AK293331.1 | - | 1 | 82 | ||
AK299511.1 | - | 1 | 120 | |||
AK307678.1 | - | 1 | 119 | |||
BC017362.2 | - | 1 | 98 | |||
BC091504.1 | - | 1 | 98 | 30379019 | ||
DA410183.1 | - | 1 | 227 | |||
L19183.1 | - | 1 | 121 |