Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284330.1 | NP_001271259.1 | 7 | 1039 | ||
NM_001284331.1 | NP_001271260.1 | 8 | 1168 | |||
NM_013284.3 | NP_037416.1 | 9 | 1279 | |||
NR_104299.1 | 9 | 1250 | ||||
XM_005249711.2 | XP_005249768.1 | 6 | 830 | |||
XM_006715692.2 | XP_006715755.1 | 9 | 1250 | |||
XM_006715696.2 | XP_006715759.1 | 7 | 980 | |||
XM_006715698.3 | XP_006715761.1 | 4 | 1134 | |||
XM_011515275.1 | XP_011513577.1 | 9 | 1265 | |||
XM_011515278.1 | XP_011513580.1 | 8 | 1115 | |||
XM_011515279.1 | XP_011513581.1 | 9 | 1265 | |||
XM_011515282.2 | XP_011513584.1 | 4 | 1131 | |||
XM_011515285.1 | XP_011513587.1 | 5 | 517 | |||
XM_017011998.1 | XP_016867487.1 | 8 | 1079 | |||
XM_017011999.1 | XP_016867488.1 | 7 | 1025 | |||
XM_017012000.1 | XP_016867489.1 | 8 | 1115 | |||
XM_017012001.1 | XP_016867490.1 | 7 | 1025 | |||
XM_017012002.1 | XP_016867491.1 | 8 | 1095 | |||
XM_017012003.1 | XP_016867492.1 | 6 | 843 | |||
GenBank mRNA | AF176097.1 | - | 9 | 1219 | ||
AJ131891.2 | - | 9 | 1222 | |||
AK023002.1 | - | 9 | 1225 | |||
AK092903.1 | - | 5 | 1002 | |||
AK307634.1 | - | 6 | 1116 | |||
BC026306.1 | - | 8 | 1822 | 4813399 | ||
BC049202.1 | - | 7 | 984 | 5246144 | ||
BC062590.1 | - | 8 | 1092 | 5215523 |