Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145076.2 | NP_001138548.1 | 20 | 2386 | ||
NM_019063.4 | NP_061936.2 | 21 | 2560 | |||
XM_005264267.1 | XP_005264324.1 | 22 | 2573 | |||
XM_006711991.1 | XP_006712054.1 | 21 | 2399 | |||
XM_006711992.2 | XP_006712055.1 | 23 | 2694 | |||
GenBank mRNA | AF177377.2 | - | 21 | 2522 | ||
AK056880.1 | - | 10 | 1062 | |||
AK295778.1 | - | 6 | 598 | |||
AK314702.1 | - | 21 | 2534 | |||
AL832392.1 | - | 7 | 713 | |||
BC140845.1 | - | 21 | 2483 | 9021713 | ||
BC146799.1 | - | 20 | 2360 | 8860304 | ||
BX647731.1 | - | 12 | 1623 |