Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163079.1 | NP_001156551.1 | 2 | 1159 | ||
NM_031890.3 | NP_114096.1 | 1 | 2223 | |||
XM_011546125.2 | XP_011544427.1 | 2 | 1569 | |||
GenBank mRNA | AF307451.1 | - | 1 | 2153 | ||
AK095437.1 | - | 1 | 26 | |||
AK095609.1 | - | 2 | 1153 |