Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290046.1 | NP_001276975.1 | 16 | 2288 | ||
NM_001290047.1 | NP_001276976.1 | 16 | 2757 | |||
XM_006724079.3 | XP_006724142.1 | 4 | 1200 | |||
XM_011546128.2 | XP_011544430.1 | 15 | 2361 | |||
XM_011546129.2 | XP_011544431.1 | 16 | 2361 | |||
XM_011546132.2 | XP_011544434.1 | 16 | 2445 | |||
XM_017028785.1 | XP_016884274.1 | 16 | 2445 | |||
GenBank mRNA | AB051527.1 | - | 8 | 1306 | ||
AF336133.1 | - | 16 | 2817 | |||
AK308330.1 | - | 16 | 2289 | |||
AL832377.1 | - | 16 | 2235 | |||
BX647449.1 | - | 16 | 2235 |