Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014510.2 | NP_055325.2 | 5 | 4953 | ||
NM_033026.5 | NP_149015.2 | 5 | 4953 |