Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005618.3 | NP_005609.3 | 3 | 834 | ||
XM_005266934.3 | XP_005266991.1 | 3 | 1300 | |||
GenBank mRNA | AF196571.1 | - | 3 | 682 | ||
AK314234.1 | - | 3 | 706 | |||
AY358892.1 | - | 3 | 401 | |||
BC028096.1 | - | 2 | 1640 | |||
DR000029.1 | - | 3 | 453 | |||
EU927387.1 | - | 3 | 525 |