Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206924.1 | NP_001193853.1 | 3 | 328 | ||
NM_001206925.1 | NP_001193854.1 | 3 | 614 | |||
NM_006889.4 | NP_008820.3 | 4 | 719 | |||
NM_175862.4 | NP_787058.4 | 4 | 664 | |||
NM_176892.1 | NP_795711.1 | 4 | 719 | |||
GenBank mRNA | AK291497.1 | - | 4 | 572 | ||
AK294663.1 | - | 3 | 324 | |||
AK301237.1 | - | 3 | 610 | |||
AK313103.1 | - | 4 | 573 | |||
AK316203.1 | - | 3 | 610 | |||
BC040261.1 | - | 4 | 652 | 5173789 | ||
CR541844.1 | - | 3 | 518 | |||
KU284848.1 | - | 3 | 518 | |||
L25259.1 | - | 4 | 642 | |||
U04343.1 | - | 4 | 665 |