Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014012.5 | NP_054731.2 | 4 | 770 | ||
XM_005260404.1 | XP_005260461.1 | 4 | 794 | |||
XM_011528795.1 | XP_011527097.1 | 4 | 788 | |||
XM_017027833.1 | XP_016883322.1 | 4 | 628 | |||
GenBank mRNA | AF084465.1 | - | 3 | 544 | ||
AF152863.1 | - | 4 | 752 | |||
AK313060.1 | - | 4 | 770 | |||
BC039813.1 | - | 4 | 756 | 5220518 | ||
BT020078.1 | - | 3 | 472 | |||
BT020079.1 | - | 3 | 472 |