Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014018.2 | NP_054737.1 | 2 | 272 | ||
GenBank mRNA | AF070663.1 | - | 2 | 256 | ||
AK026506.1 | - | 2 | 280 | |||
AK223069.1 | - | 2 | 280 | |||
AK315739.1 | - | 2 | 272 | |||
BC010150.1 | - | 2 | 272 | 4331173 | ||
KU178612.1 | - | 2 | 249 | |||
KU178613.1 | - | 2 | 264 | |||
KU178614.1 | - | 2 | 231 |