Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085454.1 | NP_001078923.1 | 7 | 953 | ||
NM_014030.3 | NP_054749.2 | 7 | 953 | |||
XM_011524684.1 | XP_011522986.1 | 7 | 794 | |||
XM_011524685.1 | XP_011522987.1 | 7 | 794 | |||
GenBank mRNA | AB209537.1 | - | 7 | 752 | ||
AF124490.1 | - | 7 | 753 | |||
AK024826.1 | - | 2 | 108 | |||
AK294785.1 | - | 8 | 831 | |||
AK299932.1 | - | 7 | 797 | |||
BC067358.1 | - | 7 | 953 | |||
BQ424386.1 | - | 6 | 592 |