Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC27A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017372.2 | NP_001017372.1 | 1 | 1262 | ||
NM_001317984.1 | NP_001304913.1 | 2 | 1086 | |||
NM_014031.4 | NP_054750.1 | 2 | 1037 | |||
GenBank mRNA | AF064254.1 | - | 1 | 404 | ||
AK313456.1 | - | 2 | 685 | |||
AL833117.1 | - | 1 | 835 | |||
BC041945.1 | - | 1 | 1261 | 5301306 | ||
BX537383.1 | - | 2 | 647 | |||
CR457129.1 | - | 1 | 251 |