Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286067.1 | NP_001272996.1 | 10 | 1223 | ||
NM_001286068.1 | NP_001272997.1 | 9 | 1086 | |||
NM_001286069.1 | NP_001272998.1 | 9 | 1091 | |||
NM_001286070.1 | NP_001272999.1 | 8 | 939 | |||
NM_001286071.1 | NP_001273000.1 | 8 | 914 | |||
NM_014039.3 | NP_054758.2 | 8 | 941 | |||
XM_006718824.1 | XP_006718887.1 | 10 | 1132 | |||
XM_011542781.2 | XP_011541083.1 | 9 | 1405 | |||
XM_011542782.2 | XP_011541084.1 | 9 | 1496 | |||
XM_017017613.1 | XP_016873102.1 | 10 | 4547 | |||
XM_017017614.1 | XP_016873103.1 | 10 | 1374 | |||
XM_017017615.1 | XP_016873104.1 | 10 | 1827 | |||
XM_017017616.1 | XP_016873105.1 | 8 | 933 | |||
XM_017017617.1 | XP_016873106.1 | 8 | 1329 | |||
XM_017017618.1 | XP_016873107.1 | 8 | 905 | |||
XM_017017619.1 | XP_016873108.1 | 7 | 787 | |||
GenBank mRNA | AF092133.1 | - | 9 | 999 | ||
AK129932.1 | - | 4 | 510 | |||
AK225977.1 | - | 9 | 1021 | |||
AK291833.1 | - | 9 | 997 | |||
AK292215.1 | - | 10 | 1142 | |||
AL136605.1 | - | 9 | 1017 | |||
AY203960.1 | - | 8 | 967 | |||
BC007110.1 | - | 8 | 846 | |||
BC012298.1 | - | 8 | 908 | 3852558 | ||
CR533538.1 | - | 8 | 859 |