Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAMDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316957.1 | NP_001303886.1 | 3 | 238 | ||
NM_001316958.1 | NP_001303887.1 | 3 | 238 | |||
NM_001316960.1 | NP_001303889.1 | 3 | 314 | |||
NM_001316961.1 | NP_001303890.1 | 3 | 234 | |||
NM_001316962.1 | NP_001303891.1 | 3 | 238 | |||
NM_024684.3 | NP_078960.1 | 2 | 196 | |||
NR_133637.1 | 2 | 196 | ||||
XM_011544968.2 | XP_011543270.1 | 2 | 196 | |||
XM_011544969.2 | XP_011543271.1 | 2 | 189 | |||
XM_017017620.1 | XP_016873109.1 | 2 | 218 | |||
GenBank mRNA | AF092136.1 | - | 3 | 197 | ||
AK024688.1 | - | 2 | 155 | |||
BC002752.2 | - | 2 | 148 | 3628061 | ||
BC016854.2 | - | 2 | 125 | 3856546 | ||
BI791786.1 | - | 3 | 247 | |||
BM930331.1 | - | 3 | 183 | |||
BP270361.1 | - | 2 | 196 | |||
BX646013.1 | - | 2 | 180 | |||
CN362204.1 | - | 3 | 209 | |||
CR457332.1 | - | 1 | 50 |