Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014050.3 | NP_054769.1 | 5 | 491 | ||
NM_172177.3 | NP_751917.1 | 5 | 494 | |||
NR_038159.1 | 6 | 514 | ||||
NR_038160.1 | 6 | 511 | ||||
NR_038161.1 | 6 | 596 | ||||
GenBank mRNA | AF078860.1 | - | 5 | 473 | ||
AF135160.1 | - | 5 | 477 | |||
AF151038.1 | - | 5 | 465 | |||
AK000285.1 | - | 5 | 467 | |||
AL136659.1 | - | 5 | 468 | |||
BC040240.2 | - | 4 | 430 | 3445213 | ||
BM147739.1 | - | 5 | 492 | |||
BM726733.1 | - | 6 | 469 | |||
BU561400.1 | - | 490 | ||||
CD385500.1 | - | 5 | 487 | |||
CR533495.1 | - | 4 | 332 | |||
CR749344.1 | - | 6 | 568 |