Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014051.3 | NP_054770.1 | 4 | 378 | ||
GenBank mRNA | AF078864.1 | - | 4 | 422 | ||
AF239771.1 | - | 4 | 357 | |||
AK312063.1 | - | 4 | 351 | |||
BC015097.1 | - | 4 | 337 | 3686196 | ||
BC019328.1 | - | 4 | 333 | 3534851 |