Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143779.1 | NP_001137251.1 | 4 | 404 | ||
NM_014055.3 | NP_054774.2 | 4 | 822 | |||
NM_031473.3 | NP_113661.2 | 4 | 822 | |||
XM_011538232.2 | XP_011536534.1 | 4 | 350 | |||
XM_017019217.1 | XP_016874706.1 | 4 | 514 | |||
XM_017019218.1 | XP_016874707.1 | 4 | 822 | |||
GenBank mRNA | AF139540.1 | - | 4 | 362 | ||
AF250326.1 | - | 3 | 315 | |||
AF332010.1 | - | 4 | 780 | |||
AK091549.1 | - | 3 | 182 | |||
AK291043.1 | - | 4 | 724 | |||
BC004536.2 | - | 4 | 658 | 3608705 | ||
BC005396.1 | - | 4 | 324 | |||
BC108257.1 | - | 3 | 287 | 6156440 | ||
BP351086.1 | - | 4 | 404 |