Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143779.1 | NP_001137251.1 | 6, 7 | 722 | ||
NM_014055.3 | NP_054774.2 | 6, 7 | 1140 | |||
NM_031473.3 | NP_113661.2 | 6, 7 | 1140 | |||
XM_011538232.2 | XP_011536534.1 | 6, 7 | 668 | |||
XM_017019217.1 | XP_016874706.1 | 6, 7 | 832 | |||
XM_017019218.1 | XP_016874707.1 | 6, 7 | 1140 | |||
GenBank mRNA | AF139540.1 | - | 6, 7 | 680 | ||
AF250326.1 | - | 5, 6 | 633 | |||
AF332010.1 | - | 6, 7 | 1098 | |||
AK091549.1 | - | 5, 6 | 500 | |||
AK291043.1 | - | 6, 7 | 1042 | |||
BC004536.2 | - | 6, 7 | 976 | 3608705 | ||
BC005396.1 | - | 6, 7 | 642 | |||
BC108257.1 | - | 5, 6 | 605 | 6156440 |