Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014061.4 | NP_054780.2 | 1 | 348 | ||
GenBank mRNA | AF143235.4 | - | 1 | 344 | ||
AF320912.1 | - | 1 | 273 | |||
AK313527.1 | - | 1 | 272 | |||
BC011954.1 | - | 1 | 299 | 3880523 | ||
BI916425.1 | - | 1 | 348 |