Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASTE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288950.1 | NP_001275879.1 | 3 | 495 | ||
NM_014065.3 | NP_054784.2 | 3 | 720 | |||
NR_110229.1 | 3 | 720 | ||||
XM_017006266.1 | XP_016861755.1 | 3 | 636 | |||
XM_017006267.1 | XP_016861756.1 | 3 | 794 | |||
XM_017006268.1 | XP_016861757.1 | 3 | 715 | |||
XM_017006269.1 | XP_016861758.1 | 3 | 636 | |||
GenBank mRNA | AK027258.1 | - | 1 | 110 | ||
AK027806.1 | - | 3 | 668 | |||
AK294156.1 | - | 3 | 720 | |||
AK301388.1 | - | 3 | 608 | |||
BC069265.1 | - | 3 | 433 | |||
BC110616.2 | - | 3 | 433 | 40033986 |