Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MACROD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014067.3 | NP_054786.2 | 9 | 1038 | ||
XM_005273939.4 | XP_005273996.1 | 9 | 1577 | |||
XM_011544970.2 | XP_011543272.1 | 8 | 1547 | |||
GenBank mRNA | AF202922.2 | - | 9 | 971 | ||
BC000270.1 | - | 9 | 986 | 3357931 | ||
BC003188.1 | - | 9 | 792 | 3502054 | ||
BC007297.1 | - | 9 | 986 | 3349763 | ||
BC008316.1 | - | 9 | 986 | 3342973 |