Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014079.3 | NP_054798.1 | 3 | 2304 | ||
GenBank mRNA | BC036733.1 | - | 3 | 2308 | 5526657 |