Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009923.1 | NP_001009923.1 | 5 | 544 | ||
NM_001009924.1 | NP_001009924.1 | 5 | 762 | |||
NM_001009925.1 | NP_001009925.1 | 4 | 438 | |||
NM_014145.4 | NP_054864.3 | 4 | 669 | |||
XM_005260713.3 | XP_005260770.1 | 4 | 636 | |||
XM_011529227.2 | XP_011527529.1 | 4 | 659 | |||
XM_011529229.2 | XP_011527531.1 | 5 | 684 | |||
XM_017027836.1 | XP_016883325.1 | 4 | 421 | |||
GenBank mRNA | AK055700.1 | - | 5 | 718 | ||
AK315606.1 | - | 4 | 428 | |||
AL357536.1 | - | 2 | 131 | |||
AY359115.1 | - | 4 | 561 | |||
BC009768.1 | - | 4 | 422 | 4092724 | ||
BC009769.1 | - | 4 | 428 | 4132400 | ||
BC009770.1 | - | 4 | 422 | 4249231 | ||
BC011990.1 | - | 4 | 409 | 3857616 | ||
BC015113.1 | - | 4 | 373 | 4049350 | ||
BC070212.1 | - | 4 | 438 | 4687557 | ||
BC110408.1 | - | 4 | 400 | 5740520 |