Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014149.3 | NP_054868.3 | 14 | 2087 | ||
XM_011516083.2 | XP_011514385.1 | 15 | 2192 | |||
XM_011516086.2 | XP_011514388.1 | 15 | 2114 | |||
XM_017012070.1 | XP_016867559.1 | 14 | 2135 | |||
XM_017012071.1 | XP_016867560.1 | 12 | 1627 | |||
XM_017012072.1 | XP_016867561.1 | 14 | 2069 | |||
GenBank mRNA | AK001606.1 | - | 14 | 2042 | ||
AK022935.1 | - | 13 | 1918 | |||
AK124765.1 | - | 1 | 2166 | |||
BC017246.2 | - | 14 | 2026 | 4890904 |