Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014149.3 | NP_054868.3 | 14 | 1980 | ||
XM_011516083.2 | XP_011514385.1 | 15 | 2085 | |||
XM_011516086.2 | XP_011514388.1 | 15 | 2007 | |||
XM_017012070.1 | XP_016867559.1 | 14 | 2028 | |||
XM_017012071.1 | XP_016867560.1 | 12 | 1520 | |||
XM_017012072.1 | XP_016867561.1 | 14 | 1962 | |||
GenBank mRNA | AF161534.1 | - | 14 | 1908 | ||
AK001606.1 | - | 14 | 1935 | |||
AK022935.1 | - | 13 | 1811 | |||
AK074466.1 | - | 7 | 740 | |||
AK124765.1 | - | 1 | 2059 | |||
AL136940.1 | - | 14 | 1831 | |||
BC009939.2 | - | 12 | 1438 | |||
BC017246.2 | - | 14 | 1919 | 4890904 | ||
BC063394.1 | - | 14 | 1916 | |||
BX640957.1 | - | 14 | 2900 | |||
CR533561.1 | - | 13 | 1762 |