Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014153.3 | NP_054872.2 | 17 | 2200 | ||
XM_006720877.2 | XP_006720940.1 | 17 | 2236 | |||
XM_011522463.1 | XP_011520765.1 | 18 | 2385 | |||
XM_011522464.2 | XP_011520766.1 | 11 | 1993 | |||
XM_017023174.1 | XP_016878663.1 | 17 | 2217 | |||
XM_017023175.1 | XP_016878664.1 | 13 | 1830 | |||
GenBank mRNA | AK000325.1 | - | 15 | 1939 | ||
AK026956.1 | - | 9 | 1425 | |||
AK091803.1 | - | 15 | 1902 | |||
AY163807.1 | - | 16 | 2229 | |||
BC046363.1 | - | 17 | 2093 | 5538820 |