Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025109.1 | NP_001020280.1 | 3 | 737 | ||
NM_001773.2 | NP_001764.1 | 3 | 737 | |||
GenBank mRNA | AF523361.1 | - | 3 | 550 | ||
AF538601.1 | - | 2 | 388 | |||
AK291529.1 | - | 3 | 490 | |||
AK293030.1 | - | 3 | 514 | |||
AK300835.1 | - | 4 | 554 | |||
BC039146.1 | - | 3 | 487 | 4746591 | ||
M81104.1 | - | 3 | 736 | |||
S53910.1 | - | 3 | 569 | |||
S53911.1 | - | 3 | 569 |