Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014165.3 | NP_054884.1 | 1, 2 | 227 | ||
GenBank mRNA | AF060508.1 | - | 1, 2 | 173 | ||
AF161474.1 | - | 1, 2 | 216 | |||
AK311850.1 | - | 1, 2 | 200 | |||
BC018802.1 | - | 1, 2 | 213 | |||
BC039464.1 | - | 1, 2 | 206 | 4388671 | ||
DB253189.1 | - | 1, 2 | 227 |
Gene Symbol: NDUFAF4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036507.1 | 1 | 162 |