Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014165.3 | NP_054884.1 | 3 | 371 | ||
GenBank mRNA | AF060508.1 | - | 3 | 317 | ||
AF161474.1 | - | 3 | 360 | |||
AK026245.1 | - | 2 | 216 | |||
AK311850.1 | - | 3 | 344 | |||
BC018802.1 | - | 3 | 357 | |||
BC039464.1 | - | 3 | 350 | 4388671 | ||
DB253189.1 | - | 3 | 371 |